ENA429358435 Enamine

2-(7-amino-1,2,3,4-tetrahydroisoquinolin-2-yl)acetamide

  • MDL number MFCD22191505
  • Cas Number : 1232787-18-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-179816

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-179816
Availability details for
Quantity
  • Availability
  • Add to Cart