ENA429358436 Enamine

2-({1,7,7-trimethylbicyclo[2.2.1]heptan-2-yl}oxy)benzaldehyde

  • MDL number MFCD22947255
  • Cas Number : 1560334-63-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-180025

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-180025
Availability details for
Quantity
  • Availability
  • Add to Cart