ENA429358441 Enamine

1-{[(tert-butoxy)carbonyl]amino}-1H-pyrrole-2-carboxamide

  • MDL number MFCD24471141
  • Cas Number : 1206824-76-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-180267

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-180267
Availability details for
Quantity
  • Availability
  • Add to Cart