ENA429358454 Enamine

2-bromo-5-propoxypyridine

  • MDL number MFCD21099946
  • Cas Number : 1144110-15-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-180711

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-180711
Availability details for
Quantity
  • Availability
  • Add to Cart