ENA429358455 Enamine

4-bromoisoquinoline-5-sulfonamide

  • MDL number MFCD28118313
  • Cas Number : 127625-93-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-180757

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-180757
Availability details for
Quantity
  • Availability
  • Add to Cart