ENA429358460 Enamine

methyl 2,2-dimethyl-3-(methylsulfanyl)propanoate

  • MDL number MFCD23731793
  • Cas Number : 876299-31-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-181507

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-181507
Availability details for
Quantity
  • Availability
  • Add to Cart