ENA429358464 Enamine

tert-butyl N-(1H-indol-6-yl)carbamate

  • MDL number MFCD04114770
  • Cas Number : 885273-73-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 106-108
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-181594

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-181594
Availability details for
Quantity
  • Availability
  • Add to Cart