ENA429358466 Enamine

2H,3H-pyrazolo[3,2-b][1,3]oxazole-6-carboxylic acid

  • MDL number MFCD16547294
  • Cas Number : 1239722-75-8
  • Molecular Weight : 154.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2,3-dihydropyrazolo[5,1-b][1,3]oxazole-6-carboxylic acid
Inchi Code 1S/C6H6N2O3/c9-6(10)4-3-5-8(7-4)1-2-11-5/h3H,1-2H2,(H,9,10)
Inchi Key VIJQAWCUVDTCAZ-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-181688

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-181688
pictograms GHS07
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart