ENA429358470 Enamine

1-(3-aminophenyl)azetidin-2-one

  • MDL number MFCD28118341
  • Cas Number : 1456595-12-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-181807

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-181807
Availability details for
Quantity
  • Availability
  • Add to Cart