ENA429358476 Enamine

2-(propan-2-yl)-4,5,6,7-tetrahydro-1H-1,3-benzodiazole-5-carboxylic acid hydrochloride

  • MDL number MFCD28118346
  • Cas Number : 1004529-30-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-181949

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-181949
Availability details for
Quantity
  • Availability
  • Add to Cart