ENA429358477 Enamine

4-chloro-N-[2-(propan-2-yl)oxan-3-yl]pyrimidin-2-amine

  • MDL number MFCD28118347
  • Cas Number : 1803571-57-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-181952

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-181952
Availability details for
Quantity
  • Availability
  • Add to Cart