ENA429358486 Enamine

1-[5-(difluoromethoxy)-4-fluoro-2-nitrophenyl]-1H-1,2,4-triazole

  • MDL number MFCD28118353
  • Cas Number : 1803571-61-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-182256

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-182256
Availability details for
Quantity
  • Availability
  • Add to Cart