ENA429358487 Enamine

hexahydro-1H-pyrrolo[2,1-c]morpholine-3-carbohydrazide

  • MDL number MFCD28118354
  • Cas Number : 1803586-84-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-182271

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-182271
Availability details for
Quantity
  • Availability
  • Add to Cart