ENA429358489 Enamine

2-methoxy-1,3-dimethyl-4-nitrobenzene

  • MDL number MFCD11036252
  • Cas Number : 50536-74-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-182279

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-182279
Availability details for
Quantity
  • Availability
  • Add to Cart