ENA429358490 Enamine

4-bromo-1,3-dimethyl-1H-pyrazole-5-carbaldehyde

  • MDL number MFCD22058061
  • Cas Number : 1367816-40-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-182333

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-182333
Availability details for
Quantity
  • Availability
  • Add to Cart