ENA429358491 Enamine

5-amino-1-(4-methylpyridin-2-yl)-3-(methylsulfanyl)-1H-pyrazole-4-carbonitrile

  • MDL number MFCD16519094
  • Cas Number : 1285256-32-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-182353

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-182353
Availability details for
Quantity
  • Availability
  • Add to Cart