ENA429358496 Enamine

3-(furan-3-yl)-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD23858155
  • Cas Number : 1513964-21-0
  • Molecular Weight : 178.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-182460

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-182460
Availability details for
Quantity
  • Availability
  • Add to Cart