ENA429358497 Enamine

6-nitro-3,4-dihydro-2H-1-benzopyran

  • MDL number MFCD15527074
  • Cas Number : 50386-60-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 99-101
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-182468

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-182468
Availability details for
Quantity
  • Availability
  • Add to Cart