ENA429358507 Enamine

2-[(5-chloro-2-nitrophenyl)amino]-3-methylbutanoic acid

  • MDL number MFCD25210235

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-182704

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-182704
Availability details for
Quantity
  • Availability
  • Add to Cart