ENA429358511 Enamine

2-(N-hydroxyimino)-N-(2,3,4-trifluorophenyl)acetamide

  • Cas Number : 1453266-30-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-182797

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-182797
Availability details for
Quantity
  • Availability
  • Add to Cart