ENA429358517 Enamine

N-(6-chloro-3-fluoro-2-methylphenyl)-2,2-dimethylpropanamide

  • MDL number MFCD28118385
  • Cas Number : 1803582-55-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-182882

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-182882
Availability details for
Quantity
  • Availability
  • Add to Cart