ENA429358524 Enamine

4-(4-chlorophenyl)-4-methylpiperidine-2,6-dione

  • MDL number MFCD18276969
  • Cas Number : 1268045-87-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-183078

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-183078
Availability details for
Quantity
  • Availability
  • Add to Cart