ENA429358527 Enamine

4-(4-chlorophenyl)-4-methyl-2,6-dioxopiperidine-3,5-dicarbonitrile

  • MDL number MFCD28118401
  • Cas Number : 92161-82-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-183081

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-183081
Availability details for
Quantity
  • Availability
  • Add to Cart