ENA429358531 Enamine

methyl 2-(1-aminoethyl)-1,3-thiazole-4-carboxylate

  • MDL number MFCD09904782
  • Cas Number : 1017469-43-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-183168

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-183168
Availability details for
Quantity
  • Availability
  • Add to Cart