ENA429358533 Enamine

2,3-dihydro-1-benzoxepin-3-one

  • MDL number MFCD24445373
  • Cas Number : 369376-68-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-183178

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-183178
Availability details for
Quantity
  • Availability
  • Add to Cart