ENA429358536 Enamine

(2-nitrophenyl)(pyridin-2-yl)methanol

  • MDL number MFCD28118418
  • Cas Number : 115177-58-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-183219

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-183219
Availability details for
Quantity
  • Availability
  • Add to Cart