ENA429358539 Enamine

methyl 2,3-dichloro-5-nitrobenzoate

  • MDL number MFCD12172998
  • Cas Number : 63105-54-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-183315

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-183315
Availability details for
Quantity
  • Availability
  • Add to Cart