ENA429358563 Enamine

(2,4-dichloro-1,3-thiazol-5-yl)methanol

  • MDL number MFCD09751262
  • Cas Number : 170232-69-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-184012

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-184012
Availability details for
Quantity
  • Availability
  • Add to Cart