ENA429358564 Enamine

5-bromo-3-[(5-ethyl-1,2,4-oxadiazol-3-yl)methyl]-2-methyl-3,4-dihydropyrimidin-4-one

  • MDL number MFCD23219750
  • Cas Number : 1502433-71-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-184033

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-184033
Availability details for
Quantity
  • Availability
  • Add to Cart