ENA429358568 Enamine

2-(4-ethyl-4H-1,2,4-triazol-3-yl)morpholine dihydrochloride

  • MDL number MFCD28118466
  • Cas Number : 1803587-59-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-184081

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-184081
Availability details for
Quantity
  • Availability
  • Add to Cart