ENA429358571 Enamine

1-(pyrimidin-5-yl)cyclohexan-1-ol

  • MDL number MFCD16779197
  • Cas Number : 1248479-24-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-184091

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-184091
Availability details for
Quantity
  • Availability
  • Add to Cart