ENA429358580 Enamine

ethyl 2-hydroxy-2-(3-methylpyridin-2-yl)acetate

  • MDL number MFCD27931009
  • Cas Number : 873459-50-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-184352

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-184352
Availability details for
Quantity
  • Availability
  • Add to Cart