ENA429358582 Enamine

4-bromo-1,3-benzothiazole-2-thiol

  • MDL number MFCD11044824
  • Cas Number : 1083181-41-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-184371

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-184371
Availability details for
Quantity
  • Availability
  • Add to Cart