ENA429358588 Enamine

1-tert-butyl-3-(furan-2-yl)-1H-pyrazol-5-amine

  • MDL number MFCD12168889
  • Cas Number : 222160-55-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-184457

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-184457
Availability details for
Quantity
  • Availability
  • Add to Cart