ENA429358589 Enamine

4-ethoxy-3-fluorobenzaldehyde

  • MDL number MFCD06247348
  • Cas Number : 452-00-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-185353

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-185353
Availability details for
Quantity
  • Availability
  • Add to Cart