ENA429358590 Enamine

4-ethoxy-3-fluorobenzonitrile

  • MDL number MFCD12110223
  • Cas Number : 1156079-46-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-185354
Availability details for
Quantity
  • Availability
  • Add to Cart