ENA429358592 Enamine

1-(benzyloxy)-2-(2-methoxyethyl)benzene

  • MDL number MFCD28118503
  • Cas Number : 1427435-95-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-185364

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-185364
Availability details for
Quantity
  • Availability
  • Add to Cart