ENA429358594 Enamine

ethyl 2,2-difluoro-3-hydroxybutanoate

  • MDL number MFCD00720836
  • Cas Number : 321942-91-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-185397

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-185397
Availability details for
Quantity
  • Availability
  • Add to Cart