ENA429358595 Enamine

1-methyl-3-(thiophen-3-yl)-1H-pyrazol-5-amine

  • MDL number MFCD11105307
  • Cas Number : 1152708-82-1
  • Molecular Weight : 179.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-185408

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-185408
Availability details for
Quantity
  • Availability
  • Add to Cart