ENA429358596 Enamine

3-chloro-1-(2,3-dihydro-1,4-benzodioxin-6-yl)propan-1-one

  • MDL number MFCD11868925
  • Cas Number : 111038-91-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-185411

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-185411
Availability details for
Quantity
  • Availability
  • Add to Cart