ENA429358598 Enamine

4-bromo-1-{1-methyl-1H-pyrazolo[3,4-d]pyrimidin-4-yl}-1H-pyrazole

  • MDL number MFCD12824398
  • Cas Number : 1178290-21-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-185420

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-185420
Availability details for
Quantity
  • Availability
  • Add to Cart