ENA429358599 Enamine

5-(chloromethyl)-3-(3-fluorophenyl)-1,2,4-oxadiazole

  • MDL number MFCD09034177
  • Cas Number : 601484-33-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-185429

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-185429
Availability details for
Quantity
  • Availability
  • Add to Cart