ENA429358602 Enamine

3-methyldodecan-1-ol

  • MDL number MFCD28118516
  • Cas Number : 91377-01-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-185513

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-185513
Availability details for
Quantity
  • Availability
  • Add to Cart