ENA429358604 Enamine

1-(difluoromethyl)-1H-pyrrole-2-carbaldehyde

  • MDL number MFCD21766137
  • Cas Number : 1519187-90-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-185535

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-185535
Availability details for
Quantity
  • Availability
  • Add to Cart