ENA429358605 Enamine

methyl 1-(2,2-difluoroethyl)-1H-pyrazole-5-carboxylate

  • MDL number MFCD09473371
  • Cas Number : 1172234-95-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-185538

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-185538
Availability details for
Quantity
  • Availability
  • Add to Cart