ENA429358607 Enamine

5-(chloromethyl)-3-[(3-fluorophenyl)methyl]-1,2,4-oxadiazole

  • MDL number MFCD14598987
  • Cas Number : 1247475-08-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-185551

Safety Information

msds https://www.enaminestore.com/catalog/EN300-185551
Availability details for
Quantity
  • Availability
  • Add to Cart