ENA429358613 Enamine

1-(difluoromethyl)-3-nitro-1H-indazole

  • MDL number MFCD28118529
  • Cas Number : 1803599-67-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-185607

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-185607
Availability details for
Quantity
  • Availability
  • Add to Cart