ENA429358614 Enamine

tert-butyl N-(2-methylpyridin-3-yl)carbamate

  • MDL number MFCD17169891
  • Cas Number : 1219095-87-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-185622

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-185622
Availability details for
Quantity
  • Availability
  • Add to Cart