ENA429358623 Enamine

3-cyclohexyl-4-(2-methylpropyl)-1,2-oxazol-5-amine

  • MDL number MFCD16068996
  • Cas Number : 1250480-94-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-185701

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-185701
Availability details for
Quantity
  • Availability
  • Add to Cart