ENA429358625 Enamine

1-methyl-4,5,6,7-tetrahydro-1H-1,2,3-benzotriazole-5-carboxylic acid

  • MDL number MFCD28118535
  • Cas Number : 1520850-00-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-185727

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-185727
Availability details for
Quantity
  • Availability
  • Add to Cart